Order Entry
Export
ContactUsLinkComponent
1,4-Thioxane ≥98%
Flammable
Irritant
1,4-Thioxane ≥98%
Catalog # A18052.06
CAS Number:  
Flammable
Irritant
1,4-Thioxane ≥98%
Catalog # A18052.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:15980-15-1
 :
Formula:C₄H₈OS
MDL Number:MFCD00006573
 :104.17 g/mol
Boiling Pt:147…148 °C
Density:1.114
Flash Pt:42 °C (107 °F)
Storage Temperature:Ambient
EINECS:240-117-6
ADR:3,III
 :GHS02
 :GHS07