Hand cleansing creams, Solopol® Light
115-1497
- Description:Hand cleansing cream, Solopol® light, 250 ml tube
- Environmentally Preferable:
- Pk:30
Solvent-free hand cleanser paste with scrubbing agent for medium- to heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust or lubricants.
- With extremely fine granulated Astopon® (refined walnut shell powder)
- Contains skin protectant Eucornol® and aloe vera
- Effective cleansing with conditioned skin feel
- Non soapy, solvent-free, colorant-free, perfumed
: EU Ecolabel
: Soft bottle may only be used with Stoko® Vario® dispensers.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904