Ladies trousers, LW97
Supplier: Portwest
Certificates
About this item
These elasticated work trousers, manufactured from the hardwearing Kingsmill fabric (65% polyester, 35% cotton), are suitable for low-hazard applications.
- Back elasticated waist allows unrestricted movement
- Sewn in crease gives the garment a sharp, tailored look
Weight: 210 g/m²
Specifications
- Catalog No:
- 113-7457
- 113-7450
- 113-7454
- 113-7449
- 113-7462
- 113-7461
- 113-7460
- 113-7456
- 113-7453
- 113-7455
- 113-7451
- 113-7459
- 113-7458
- 113-7452
- Model:
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- LW97
- Garment application:
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- Weight:
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²