Orders LinkContactUsLinkComponent
Digestion tubes, plain neck
Digestion tubes, plain neck
Supplier:   SciLabware
Digestion tubes, plain neck
Supplier:  SciLabware
Certificates
Digestion tubes, plain neck
Digestion tubes, plain neck
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Designed for use with block heaters.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Socket size
    Availability
    Price
  • 100 ml
    300 mm
    26 mm
    - NS
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 123,00 €
    123,00 €
    1 * 5 items
       123,00 €
  • 250 ml
    300 mm
    42 mm
    - NS
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 196,00 €
    196,00 €
    1 * 5 items
       196,00 €