Chemical resistant overalls, 4540+
Supplier: 3M
Certificates
About this item
These disposable overalls are made of a high quality laminated material and provide a barrier protection against light liquid splashes and hazardous dusts.
- Elasticated waist and ankles for freedom of movement and knitted cuffs for extra comfort
- Breathable back panel (SMMMS) for reduced heat stress and increased comfort
- Three-panel hood design for a better fit and compatibility with other PPE
- Two-way zip and sealable storm flap for enhanced protection
- Low-linting properties and anti-static coating
Typical applications may include paint spraying, industrial and pharmaceutical processing, asbestos inspection, insulation laying, and vehicle and machine maintenance (oils and greases).
Colour: White/blue
Certifications: Type 5/6, EN 1149-5, EN 1073-2:2002; Cat. III.
Specifications
- Catalog No:
- 113-6307
- 113-6306
- 113-6301
- 113-6305
- 113-6303
- 113-6304
- Model:
- 4540+
- 4540+
- 4540+
- 4540+
- 4540+
- 4540+
- Anti-static:
- +
- +
- +
- +
- +
- +
- Garment application:
- General Purpose, Medical, Laboratory, Industrial
- General Purpose, Medical, Laboratory, Industrial
- General Purpose, Medical, Laboratory, Industrial
- General Purpose, Medical, Laboratory, Industrial
- General Purpose, Medical, Laboratory, Industrial
- General Purpose, Medical, Laboratory, Industrial