Cut resistant gloves, Krytech 586
Supplier: MAPA
Certificates
About this item
This high performance glove, made of PU with a seamless textile support from HDPE fibres, provides protection for intricate handling in reasonably clean environments.
- HDPE fibres ensure excellent cut resistance (level 5) with reduced thickness for good dexterity
- Ventilated back for breathability
- Visible indicator in blue textile to easily identify the cut-performance level in the workplace
Suitable when handling sharp objects and large metal plates, for example in the automotive or mechanical industry.
Certifications: EN 388 (4X43D); Cat. II
Specifications
- Catalog No:
- 112-2814
- 112-2817
- 112-2816
- 112-2818
- 112-2813
- 112-2815
- Model:
- Krytech 586
- Krytech 586
- Krytech 586
- Krytech 586
- Krytech 586
- Krytech 586
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HDPE
- HDPE
- HDPE
- HDPE
- HDPE
- HDPE
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (4X43D)
- EN 388 (4X43D)
- EN 388 (4X43D)
- EN 388 (4X43D)
- EN 388 (4X43D)
- EN 388 (4X43D)