N-(2,4-Dimethoxyphenyl)-2-chloroacetamide
Catalog # MAYBBTB06598.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:101908-41-2
Formula:C₁₀H₁₂ClNO₃