(+)-Cinchonine
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
- CAS Number:118-10-5
- Formula:C₁₉H₂₂N₂O
- MDL Number:MFCD00064372
- Molecular Weight:294.4 g/mol
- Merck Index:13,02308
- Melting Pt:261…264 °C
- Density:1.2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-234-6
2 Options Available Below
Product Details & Documents
Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
- CAS Number:118-10-5
- Formula:C₁₉H₂₂N₂O
- MDL Number:MFCD00064372
- Molecular Weight:294.4 g/mol
- Merck Index:13,02308
- Melting Pt:261…264 °C
- Density:1.2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-234-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



