Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium alginate

Sodium alginate

Supplier:   Spectrum Chemical
Sodium alginate
Supplier:  Spectrum Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Alginic acid sodium salt, Algin
  • CAS Number:9005-38-3
  • MDL Number:MFCD00081310
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 210,00 €
    210,00 €
    1 * 500 g
       210,00 €

  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 797,00 €
    797,00 €
    1 * 2,5 kg
       797,00 €

  • Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg

    Specifications:
    New Product
    New Product