Phenolphthalein ACS
: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
:77-09-8
:
:C₂₀H₁₄O₄
:MFCD00005913
:1.3 g/cm³ (20 °C)
:318.33 g/mol
:258…262 °C
:14,07243
:201-004-7
:01-2119498295-24
:GHS08
:Ambient
- Environmentally Preferable:
- Clarity of Alcohol Solution:Pass
- Visual Transition Interval (pH 8 – 10):Pass