2,2-Bis(4-chlorophenyl)ethanol
Catalog # EHERC12070000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:267.15 g/mol
EINECS:220-148-1
CAS Number:2642-82-2
Formula:C₁₄H₁₂Cl₂O