1,2,3,4,6,7-Hexabromonaphthalene
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:75625-24-0
- Formula:C₁₀H₂Br₆
2 Options Available Below
Product Details & Documents
- CAS Number:75625-24-0
- Formula:C₁₀H₂Br₆
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



