Rhodium(III) oxide pentahydrate ≥99.99% (metals basis) Rh ≥ 59%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39373-27-8
:
Formula:O₃Rh₂·5H₂O
MDL Number:MFCD00149843
:343.88 g/mol (253.81 anhy)
Melting Pt: Dec.
Storage Temperature:Ambient
EINECS:234-846-9