Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2,4-difluorotoluene
5-Bromo-2,4-difluorotoluene ≥98%
5-Bromo-2,4-difluorotoluene ≥98%
Irritant
5-Bromo-2,4-difluorotoluene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
5-Bromo-2,4-difluorotoluene ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
159277-47-1
:
Formula:
C₇H₅BrF₂
MDL Number:
MFCD15144620
:
207.02 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAH32155
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation