Isobutyl-4-hydroxybenzoate
Glass bottle for solids, 100 mg
Catalog # PROOMM0496.05
Supplier: LGC Standards PROMOCHEM
Isobutyl-4-hydroxybenzoate
Catalog # PROOMM0496.05
Supplier: LGC Standards PROMOCHEM
CAS Number: 4247-02-3
Jump to:
Product Details & Documents
Synonyms: Isobutyl 4-hydroxybenzoate, Isobutylparaben
- CAS Number:4247-02-3
- Formula:C₁₁H₁₄O₃
- MDL Number:MFCD00020167
- Molecular Weight:194.23 g/mol
- EINECS:224-208-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



