Order Entry
Export
ContactUsLinkComponent
4,6-Dimethoxysalicylaldehyde
4,6-Dimethoxysalicylaldehyde
Catalog # MOLE67209584-5G
Supplier:  Molekula
CAS Number:  
4,6-Dimethoxysalicylaldehyde
Catalog # MOLE67209584-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :182.1733 g/mol
EINECS:211-904-1
CAS Number:708-76-9
Formula:C₉H₁₀O₄
MDL Number:MFCD00003328