Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichloro-3-thiophenesulphonyl chloride
2,5-Dichloro-3-thiophenesulphonyl chloride ≥97%
2,5-Dichloro-3-thiophenesulphonyl chloride ≥97%
:
Thermo Fisher Scientific
2,5-Dichloro-3-thiophenesulphonyl chloride ≥97%
:
Thermo Fisher Scientific
:
56946-83-9
:
:
C₄HCl₃O₂S₂
:
MFCD00051665
:
251.54 g/mol
:
256…257 °C
:
1.697
:
>110 °C (230 °F)
:
Ambient
:
3265
:
8,II
:
GHS05
msds_ALFAL06461
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years