Methyl-3,3-dimethoxypropionate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7424-91-1
:
Formula:C₆H₁₂O₄
MDL Number:MFCD00010650
:148.16 g/mol
Boiling Pt:76…77 °C (20 mmHg)
Density:1.06
Flash Pt:65 °C (149 °F)
Storage Temperature:Ambient
EINECS:231-055-0