Order Entry
Export
ContactUsLinkComponent
3,5-Difluorosalicylic acid ≥98%
Irritant
3,5-Difluorosalicylic acid ≥98%
Catalog # H32940.03
CAS Number:  
Irritant
3,5-Difluorosalicylic acid ≥98%
Catalog # H32940.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:84376-20-5
 :
Formula:C₇H₄F₂O₃
MDL Number:MFCD06203609
 :174.10 g/mol
Melting Pt:187…191 °C
 :GHS07
Storage Temperature:Ambient