Order Entry
Export
ContactUsLinkComponent
Rhodium(II) acetate dimer dihydrate ca 43% Rh
Irritant
Rhodium(II) acetate dimer dihydrate ca 43% Rh
Certificates
undefined
Irritant
Rhodium(II) acetate dimer dihydrate ca 43% Rh

Specification Test Results

  • Appearance:
    Green Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    43 % (Rh)

Specification Test Results

Related Information