Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhodium (II) acetate dimer
Rhodium(II) acetate dimer, Sigma-Aldrich®
Rhodium(II) acetate dimer, Sigma-Aldrich®
Rhodium(II) acetate dimer, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Rhodium(II) acetate dimer, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
15956-28-2
:
Formula:
C₈H₁₂O₈Rh₂
MDL Number:
MFCD00003538
:
M-Clarity™ quality level = MQ200
:
441.987 g/mol
EINECS:
240-084-8