Order Entry
Export
ContactUsLinkComponent
2-Mercaptopyridine-N-oxide sodium salt, Sigma-Aldrich®
2-Mercaptopyridine-N-oxide sodium salt, Sigma-Aldrich®
Catalog # SIAL329061-5G
Supplier:  Merck
CAS Number:  
2-Mercaptopyridine-N-oxide sodium salt, Sigma-Aldrich®
Catalog # SIAL329061-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :149.15 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:223-296-5
CAS Number:3811-73-2
Formula:C₅H₄NNaOS
MDL Number:MFCD01941547