2,7-Bis(bromomethyl)naphthalene, Sigma-Aldrich®
Catalog # SIAL53658-1G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:314.158 g/mol
CAS Number:38309-89-6
Formula:C₁₂H₁₀Br₂
MDL Number:MFCD08277010