Molybdenum disilicide, Sigma-Aldrich®
Catalog # SIAL243647-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:152.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:235-231-8
CAS Number:12136-78-6
Formula:MoSi₂
MDL Number:MFCD00151535