Order Entry
Export
ContactUsLinkComponent
Tris(trimethylsilyl)borate
Tris(trimethylsilyl)borate
Certificates
undefined
Tris(trimethylsilyl)borate

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3850 to 1.3870 (20°C, 589 nm)
  • GC:
    ≥97.0 %

Specification Test Results

Related Information