Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-[(Trimethylsilyl)ethynyl]benzaldehyde
4-[(Trimethylsilyl)ethynyl]benzaldehyde ≥97%
4-[(Trimethylsilyl)ethynyl]benzaldehyde ≥97%
Irritant
4-[(Trimethylsilyl)ethynyl]benzaldehyde ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
4-[(Trimethylsilyl)ethynyl]benzaldehyde ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
77123-57-0
:
Formula:
C₁₂H₁₄OSi
MDL Number:
MFCD02093765
:
202.33 g/mol
Melting Pt:
66…70 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH55060
Videos
Thermo Scientific Acroseal 25 Years
Documentation