3,3-Diethoxy-1-propylboronic acid pinacol ester ≥97%
Catalog # L19532.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:165904-27-8
:
Formula:C₁₃H₂₇BO₄
MDL Number:MFCD03788723
:258.17 g/mol
Storage Temperature:Ambient