


1,2:7,8-Diepoxyoctane ≥97%
Catalog # L06768.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
CAS Number:2426-07-5
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00005155
:142.20 g/mol
Boiling Pt:238…240 °C
Density:0.997
Flash Pt:97 °C (206 °F)
EINECS:219-375-9
ADR:8,III
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient