Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethyl-5-thiazolecarboxylic acid
2,4-Dimethyl-5-thiazolecarboxylic acid
2,4-Dimethyl-5-thiazolecarboxylic acid
2,4-Dimethyl-5-thiazolecarboxylic acid
Supplier:
Thermo Scientific
Certificates
Expand image
2,4-Dimethyl-5-thiazolecarboxylic acid
Supplier:
Thermo Scientific
About this item
CAS Number:
53137-27-2
:
Formula:
C₆H₇NO₂S
MDL Number:
MFCD00052943
:
157.1900 g/mol