
3-Bromo-1-(trimethylsilyl)-1-propyne ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38002-45-8
:
Formula:C₆H₁₁BrSi
MDL Number:MFCD00134460
:191.14 g/mol
Boiling Pt:44…45 °C (2 mmHg)
Density:1.17
Flash Pt:63 °C (145 °F)
:GHS07
Storage Temperature:Ambient