1,2-Epoxycyclooctane, Sigma-Aldrich®
Catalog # SIALC110507-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:126.196 g/mol
ADR:4.1,II
:M-Clarity™ quality level = MQ100
EINECS:206-010-3
UN:1325
CAS Number:286-62-4
Formula:C₈H₁₄O
MDL Number:MFCD00005164