Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Iodo-2,4-dimethoxypyrimidine
5-Iodo-2,4-dimethoxypyrimidine ≥98%
5-Iodo-2,4-dimethoxypyrimidine ≥98%
Irritant
5-Iodo-2,4-dimethoxypyrimidine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
5-Iodo-2,4-dimethoxypyrimidine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
52522-99-3
:
Formula:
C₆H₇IN₂O₂
MDL Number:
MFCD00090865
:
266.04 g/mol
Melting Pt:
71…75 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB24967
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation