

Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1458-18-0
:
Formula:C₆H₅Cl₂N₃O₂
MDL Number:MFCD00010431
:222.03 g/mol
Melting Pt:227…230 °C
Storage Temperature:Ambient
EINECS:215-947-7
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Light brown to red-brown Powder
- Infrared spectrum:Conforms
- Melting point:229 to 235 °C (Dec)
- HPLC:≥97.5 %