
3-Bromo-1,8-naphthalic anhydride ≥97%
Catalog # L17683.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:24050-49-5
:
Formula:C₁₂H₅BrO₃
MDL Number:MFCD00228162
:277.08 g/mol
Melting Pt:244…246 °C
:GHS07
Storage Temperature:Ambient