Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Anthraquinone
1,4-Anthraquinone ≥94%
1,4-Anthraquinone ≥94%
1,4-Anthraquinone ≥94%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
1,4-Anthraquinone ≥94%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
635-12-1
:
Formula:
C₁₄H₈O₂
MDL Number:
MFCD00013724
:
208.22 g/mol
Melting Pt:
∼ 218 °C (dec.)
Storage Temperature:
Ambient
EINECS:
211-228-7
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL00573
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation