Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyclohexyldiphenylphosphine
Cyclohexyldiphenylphosphine, Sigma-Aldrich®
Cyclohexyldiphenylphosphine, Sigma-Aldrich®
SIAL510742-5G
:
Merck
:
Cyclohexyldiphenylphosphine, Sigma-Aldrich®
SIAL510742-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
268.33 g/mol
:
M-Clarity™ quality level = MQ100
:
228-904-2
:
6372-42-5
:
C₁₈H₂₁P
:
MFCD00046360