Estriol
Glass bottle for solids, 1 g
Catalog # FITZ30-AE31
Supplier: FITZGERALD INDUSTRIES INTL
Jump to:
Product Details & Documents
- CAS Number:50-27-1
- Formula:C₁₈H₂₄O₃
- MDL Number:MFCD00003691
- EINECS:200-022-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



