Order Entry
Export
ContactUsLinkComponent
Pesticide reference standard, Chlordane 50 µg/ml in acetone
Pesticide reference standard, Chlordane 50 µg/ml in acetone
Catalog # SIALNC41805-4X1ML
Supplier:  Merck
CAS Number:  
Pesticide reference standard, Chlordane 50 µg/ml in acetone
Catalog # SIALNC41805-4X1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 mL
  • CAS No.:
    57-74-9
  • Description:
    Chlordane 50 µg/ml in acetone
  • Formula:
    C₁₀H₆Cl₈
  • Description:
    Chlordane

Specifications

About this item