2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture), Sigma-Aldrich®
Catalog # SIAL278386-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:226.3550 g/mol
:M-Clarity™ quality level = MQ100
EINECS:204-809-1
CAS Number:126-86-3
Formula:C₁₄H₂₆O₂
MDL Number:MFCD00008942