Orders LinkContactUsLinkComponent
wheaton_vials_group_900.jpg
Vials for Every Need
Wheaton Vials for Every Need | Avantor
Product selection
wheaton_scintillation_90.jpg
WHEATON™ Scintillation Vials are designed for compatibility with all scintillation counters that accommodate 20mL vials. Choose from a variety of vial, cap and liner materials. Vial materials include glass, HDPE and PET. Vials are available with caps atta
wheaton_sample_vials_90.jpg
WHEATON offers a comprehensive line of sample vials and caps for storage and chemical compatibility with a wide variety of sample types. Choose from a range of sizes and Type I clear or amber glass.
wheaton_serum_bottles_90.jpg
Tubular design provides excellent clarity and dimensional consistency from vial to vial. Specially designed bottom radius adds strength for lyophilization applications. Vials are autoclavable. Clear vials manufactured from low extractable borosilicate gl
wheaton_dualfusion_90.jpg
Combines best properties of both plastic and glass into one vial