Orders LinkContactUsLinkComponent
WHEATON.jpg
Bottles for every lab
No matter the application, WHEATON has the right bottle for you
Wheaton Bottles | Avantor
Product selection
wheaton_media_bottles_grp_90.jpg
WHEATON™ Media Bottles are a mainstay in laboratories across the world. All WHEATON Media Bottles with caps feature sloping shoulders to facilitate pouring and attached pour rings to eliminate dripping. 
wheaton_serum_bottles_90.jpg
WHEATON Serum Bottles are offered in sizes that range from 2mL to 500mL. They are made from borosilicate glass that conforms to USP Type I requirements.
wheaton_plastic_bottles_90.jpg
Leak-resistant, HDPE, LDPE, PP and much more.
wheaton_duran_bottles_90.jpg
As one of the world‘s top borosilicate glass, Duran® glass has proven itself excptional in both laboratories as well as in industrial applications.