ContactUsLinkComponent
heroblockimage955x590.jpg
Tuttnauer laboratory autoclaves, D-Line Tuttnauer
2840EL-D _ 2840ELC-G-D540x390.jpg
Front loading (horizontal)
Essential EL-D
  • Easy to clean chamber
  • Manual filling and draining of the DEMI-water reservoir
  • Re-use of the DEMI water
High end ELC-G-D
  • Full utilities are required – tap and DEMI water (not 2840), drain connection, compressed air and electricity
  • Automatic filling and draining functionality
  • Automatic cooling of the exhaust water to drain at 60 °C
Autoclaves, front loading, D-Line
2840ELV-D540x390.jpg
Top loading (vertical)
Essential ELV-D
  • Automatic filling and draining of the chamber
  • Automatic cooling of the exhaust water to drain at 60 °C
  • AISI 304 stainless steel housing
  • Essentially 'stand-alone' ELV-WR-D with water reservoir - only needs power connection.
High end ELVC-G-D
  • Full utilities are required – tap and DEMI water (not 2840), drain connection, compressed air and electricity
  • Automatic filling and draining functionality
  • Automatic cooling of the exhaust water to drain at 60 °C
  • AISI 304 stainless steel housing
Autoclaves, top loading, D-Line
undefined
Tuttnauer
Tuttnauer has been manufacturing sterilisers for over 90 years. As a global leader in sterilisation and infection control solutions, the Tuttnauer autoclaves are trusted by hospitals, research institutes, clinics, and laboratories in more than 140 countries.
Request call/demo
Tuttnauer Autoclaves: overview
Tuttnauer’s laboratory autoclaves cover a wide range of applications suitable for many fields like research institutes, universities, pharmaceutical, food, medical and biotechnological industries etc.
The advanced laboratory line is available in an unmatched range of reliable bench top (front loading) and vertical (top loading) models with chamber volumes from 23 to 160 litres, and provides a single solution for the full spectrum of sterilisation needs.
The autoclaves can be fully configured from general use to customer specific.
General specification for Laboratory autoclaves
  • High quality chamber with mirror finished door
  • Advanced control system with easy to use 3-button keyboard
  • Up to 20 customisable cycle programs
  • Standard USB and Ethernet connection for easy transfer of cycle data to PC
  • For Lab use: 2 flexible PT100 temperature sensors
Note: All 2840 and basic 3850 models require single-phase supplies. High-end 3850, all 3870, 5050 and 5075 models require a 400 volt, 3-phase supply.
Tuttnauer Autoclaves options: All Information
Each model can be upgraded with a number of options;
  • Factory fitted options: need to be ordered with the autoclave
  • General options
  • Documentation options: Innovative option is R.PC.R Reporting (Remote PC Reporting)
Download the pdf for option details
Tuttnauer Loading options: Baskets - shelves - loading tools- tables
Tuttnauer offers a broad range of loading accessories.
  • Stainless steel wire baskets and closed containers in different sizes for all autoclave models.
  • Lifting Devices for easy loading and unloading of heavy items. The lifting device is mobile or attached to the autoclave and is equipped with an integrated swivel arm for maximum maneuverability. It is also equipped with an electronic remote control for smooth handling of all load types.
  • Loading Equipment: Loading carts and transfer carriages on rails to assist the loading and unloading process. Constructed of high quality, durable stainless steel. The adjustable loading cart rolls from the transfer carriage onto the interior chamber tracks for easy handling of heavy loads.
Download the pdf for more information.
Info on services & IQOQ