ContactUsLinkComponent
Hero_SF_1842.jpg
Performance level 2: High-throughput analysis, When robustness and reproducibility are key
Be compliant-ready for your high-throughput samples in regulated environments
Choose Thermo Scientific SureSTART Performance level 2 products, to ensure high quality data with an uninterrupted workflow in high throughput applications using GC, HPLC/UHPLC, single or triple quadrupole MS systems. Use these Level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. These products have very low metal content and extractables and leachables and conform to high standards so that you can meet your regulatory requirements. They are Specification Certified to ensure their physical dimensions and autosampler compatibility.
If you need products that support routine analyses, our Performance Level 2 products are here for you. Select the products with the two waves.
Within Performance Level 2 you will find a selection of kits (including our range of EPA and Scintillation), vials, caps,inserts, septa and stoppers.
  • All products within this performance level are suitable for GC, HPLC/UHPLC, and mass spectrometry (MS) applications
  • Volume sizes ranging from 0.3 mL to 20 mL covering limited sample volumes to headspace analyses
Specification certified
  • All Thermo Scientific SureSTART vials are dimensionally verified to ensure autosampler compatibility
  • Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances
  • Vials meet all requirements of US, EU, JPN Pharmacopeia
  • Uniform and defect-free
Lot traceable
  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request
Performance level 2 products include:
SureSTART Vials/ Closures 
Glass: Amber / Clear
Inserts
Headspace Glass: Amber/Clear
Vial/Cap. Septa Kits
Crimpers, Decrimpers and Decapping
SureSTART Well Plates, Mats, Sealing Tapes, Handheld Applicators