Life Science Chromatography Products
Innovative products from sample preparation to sample handling and sepraration allowing confident identification and characterization of samples in your life science workflow.
Product selection
Thermo Scientific™ SOLA™ products provide unparalleled performance characteristics for bioanalysis compared to conventional SPE due to the innovative fritless design...
Achieve ultrahigh resolution and high efficiency separations of proteins and glycoproteins, monoclonal antibodies, biosimilars, and charge variants.
DNAPac™ PA200 AND PA200 RS (Rapid Separation UHLC) anion exchange columns are designed specifically to provide fast, high-resolution separations of single stranded nucleic acids.
MAbPac™ RP LC Columns
For high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.
MAbPac™ SCX-10 LC Columns
Used for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue.
MAbPac™ SEC-1 Columns
For high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis.
MAbPac™ HIC-10 Columns
A high resolution silica column designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction.
MAbPac™ HIC-20 HPLC Columns
Used for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity.
MAbPac™ HIC-Butyl HPLC Columns
Use to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups.
A high-performance, silica-based chromatography column that achieves mixed-mode separation using a combination of charge, isomer structure, and size.
DNAPac™ RP Columns
Achieve superior reversed phase oligonucleotide separations. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS.