ContactUsLinkComponent
lq.jpg
Automation Tips for Liquid Handling Systems
With hundreds of Thermo Scientific™ automation tips designed to fit over 50 workstations and pipetting head variations, we're certain you will find tips that increase efficiency through accurate...
With hundreds of Thermo Scientific™ automation tips designed to fit over 50 workstations and pipetting head variations, we're certain you will find tips that increase efficiency through accurate and precise performance.
These tips undergo a rigorous quality control program to ensure consistency resulting in superior tip straightness and a low coefficient of variation ranging from 1% to 3%. With stringent manufacturing specifications, our tips adhere to tolerances 400% tighter than standard pipette tips.
Tecan-Automated.jpg
  • Conductive option provides increased pipetting performance
  • Tips are validated on the actual workstations for which they are created
  • 20 μl tips increase precision of low volume pipetting through minimization of dead air space
Tecan Automation Tips
Tips-PerkinElmer-540.jpg
  • Conductive option provides increased pipetting performance
  • Increased precision for your toughest multi-dispensing assays
  • Tips are validated on the actual workstations for which they are created
PerkinElmer/Packard Robotic Automation Tips
Tips-Agilent.jpg
  • Low percentage coefficient of variation
  • Available in nonsterile, sterile or sterile filtered
  • Validated on the actual workstation for which the tip is created
Agilent Automation Tips
Tips-Eppendorf-540.jpg
  • Low percentage coefficient of variation
  • Available in nonsterile, sterile or sterile filtered
  • Validated on the actual workstation for which the tip is created
Eppendorf Automation Tips
Automated-Liquid-Handling.jpg
  • Qiagen Universal Systems
  • Molecular Devices Systems
  • Caliper/Zymark Liquid Handling Workstations
  • BioTek Pipetting Systems
  • Thermo Scientific Automated Liquid Handling Platforms
Other Automation Tips