ContactUsLinkComponent
BactronAnaerobic_Header.jpg
BACTRON Anaerobic Chambers
BACTRON chambers are total anaerobic process workstations. Samples can be prepared, cultured, and inspected in the oxygen-free chamber and standard incubator. Unlike anaerobic jars and bags, which unavoidably expose samples to oxygen, BACTRON units provide complete anaerobic environments for all your applications.
  • Ergonomic arm ports provide a comfortable work space
  • Integraded vacuum pump for quieter operation
  • 60% faster airlock cycle with 3,4 or 5 auto cycle iterations
  • Auto-sleeve cycle with integrated HEPA filter, 40% faster sleeve cycle
  • Low gas supply audible and visual alarm
  • Single button auto-commissioning
BactronAnaerobic_image1.jpg
BACTRON300 delivers optimum productivity in a compact space.
BactronAnaerobic_image2.jpg
BACTRON chambers allow efficient and dexterous, glove-free handling and inspection of samples.
BactronAnaerobic_image3.jpg
With anaerobic chambers, productivity is further improved by an up to 50% reduction in sample turnaround time.
BactronAnaerobic_image4.jpg
The BactronEZ is an ideal first anaerobic chamber or replacement chamber for users who desire to upgrade their existing chamber.