Orders LinkContactUsLinkComponent
qScript-dna-hero.jpg
qScript® cDNA Synthesis Kits
qScript cDNA synthesis kits use advanced reverse transcriptase technology for maximum yield and sensitivity for low-abundance RNA detection.
qScript® cDNA Synthesis Kits | Avantor
Our advanced reverse transcriptase technology is synonymous with maximum yield and sensitivity. qScript first-strand cDNA synthesis reagents are rigorously optimized to provide sensitive and reliable detection of low-abundance RNA for qPCR assays. A broad linear dynamic range of input RNA provides reliable assay sensitivity for robust gene expression analysis.
cDNA-SuperMix-400.jpg
High-performance qScript RT pre-blended with ribonuclease-inhibitor protein and optimized primer blend, ensuring integrity of low-abundance RNA and unbiased representation.
XLT-cDNA-400.jpg
Next-gen qScript XLT with novel hot start RT technology in an easy-to-use 5x master mix for superior 2-step RT-qPCR assay performance and reproducibility.
XLT-One-400.jpg
qScript XLT 1-Step RT-qPCR ToughMix
A highly sensitive, ready-to-use master mix for quantitative reverse transcription PCR (RT-qPCR) of RNA templates using probe-based detection methods.