Orders LinkContactUsLinkComponent
Qorpaknew.jpg
Qorpak
Qorpak - Your Source for Innovative Laboratory Packaging
Qorpak, a division of Berlin Packaging, is a leading worldwide supplier of laboratory containers and supplies. For over 40 years, Qorpak has served laboratory research and testing professionals through distributor channels in an effort to provide customers with the widest breadth of options for their needs.
Qorpak | Avantor
The right choices in laboratory packaging can make a dramatic difference in the efficiency and effectiveness of research and testing. It is important to consider your product and application when choosing the perfect bottle, closure or bottle and closure combination. Qorpak provides laboratory packaging options that deliver the solutions you need for success.
Product selection
Clear-Glass.jpg
Space-saving and easy to fill, French Squares hold the maximum volume of contents while taking up the minimum shelf and lab bench storage.
Amber.jpg
Wide Mouth Amber Straight Sided Rounds provide UV protection and offer maximum storage capacity.
Standard.jpg
Standard Clear Wide Mouth Bottles
These ideal general-purpose bottles feature a wide opening, beneficial for many applications.
Clear-Stright.jpg
Clear Straight Sided Rounds offer maximum storage capacity and optimal content visability.