Orders LinkContactUsLinkComponent
ohaus-supplier-955.png
More Than a Century of Precision and Reliability
When your work relies on consistent results, you need equipment you can trust, day after day, project after project. Trust OHAUS to deliver consistent performance. The brand is committed to reliability.
OHAUS | Avantor
For more than a century, the OHAUS name has been synonymous with precision and reliability. High-quality balances and scales, laboratory equipment and analytical instruments are designed to meet the evolving needs of a growing range of industries. OHAUS maintains an unwavering dedication to four principles that guide the way business is conducted and customers are supported: Trust, Agility, Ingenuity and Commitment. This is known as the OHAUS Advantage, a promise made to customers every day.
Balances
Discover a wide range of balances from portable to analytical and precision to semi-micro, covering all needs for weighing in every lab.
Explorer-Plus™ balances
Explorer™ and Explorer Plus™ balances
The new Explorer series offers a premium weighing performance, compliance support and data traceability. This Flagship balance is crafted to meet the most demanding requirements of modern scientific research.
Adventurer-balances-400.jpg
Adventurer® balances
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities.
Pioneer-400.jpg
Pioneer PX® balances
The OHAUS Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
Portable-400.jpg
Portable Balances
OHAUS offers a range of precision portable balances for various environments, such as the Navigator for harsh industrial settings or the Scout series for the classroom.
Moisture analysis
The OHAUS MB series Moisture Analyzers have a sleek design and offer improved durability and stability in performance, with an upgraded loadcell and carbon fiber heating technology.
MB120-400.jpg
The MB120 has the capability to produce fast and accurate results due to features such as a halogen heating system, customizable shut-off criteria and four built-in drying profiles. Get started quickly with SmartGuide™. This revolutionary feature automatically analyzes a sample and creates a method, thereby removing the guesswork from the initial setup.
MB92-400.jpg
High Performance with Advanced Features for the Most Demanding Applications. Robust and durable with the new carbon heating technology and an upgraded full-color capacitive touchscreen, MB92 is the perfect choice for users demanding a high level of performance and precision.
MB62-400.jpg
Durability and Flexible Performance in a Variety of Moisture Applications. Designed with durability and simplicity in mind, the wide 5-inch LCD display, knob control and new carbon fiber heating technology guarantee a longer service life and reliable results.
MB32-495.jpg
Perfect for Routine Moisture Testing. An LCD with second line display, navigation knob and intuitive operation system makes for convenient routine moisture testing.
MB32-400.jpg
Outstanding Performance, Flexible Configuration and Reliable Results. Halogen heating and dual weight-temperature calibration provide fast, reliable results.
Need help in choosing the right balance