Orders LinkContactUsLinkComponent
Micronova-featured.jpg
Micronova - Contamination Control Made Simpler
Micronova’s unique portfolio of products is designed to simplify contamination control and prevention. 
Micronova | Avantor
multi-surface.jpg
Cleanroom Mops in flat or
T styles that are ideal for cleaning room and equipment surfaces in controlled environments and aseptic areas.
floor-string.jpg
Cleanroom String Mops in economical and absorbent materials are well suited for cleaning large production and lab floors in a number of industries.
buckets.jpg
Buckets, carts, and wringers for flat head and string mop styles are designed to ensure proper disinfectant or detergent application.
bag_liner.jpg
Liners for storage of raw materials, disinfectant containment, waste disposal, and biohazardous material containment are available in a range of configurations for controlled environments and aseptic areas.
speciality_cleaning.jpg
Specifically designed tools meet the challenges of cleaning where equipment or surfaces are contoured, have small clearances, or are hard to reach.
detergent.jpg
Non-ionic detergents are formulated to remove particles and residues common in cleanroom and critical laboratory spaces.
cleanroom-tapes.jpg
A range of tapes accommodate applications including general marking, sealing, safety, autoclave indication, extreme temperatures, and sterile environments.
cleanroom-pens.jpg
Clear documentation is critical in regulated and controlled environments. Low sodium or gamma irradiated cleanroom pens, irradiated Sharpie® markers, IPA-resistant LabMarker™, and rollerball gel pens are among the choices for the laboratory or production.
brushes.jpg
Brushes are ideal for cleaning both small parts, such as tubing, fittings, and large overhead pipes, and drains. Micronova's robust product offering has a brush to fit all your needs.